792624
6,12-Bis(2,3,4,5,6-pentafluorophenyl)indeno[1,2-b]fluorene
95%
Synonym(s):
di-C6F5-IF
About This Item
assay
95%
form
powder
mp
380-385 °C
SMILES string
FC1=C(F)C(F)=C(F)C(F)=C1C2=C3C=CC=CC3=C4C=C5C(C6=C(F)C(F)=C(F)C(F)=C6F)=C7C=CC=CC7=C5C=C42
InChI
1S/C32H10F10/c33-23-21(24(34)28(38)31(41)27(23)37)19-13-7-3-1-5-11(13)15-9-18-16(10-17(15)19)12-6-2-4-8-14(12)20(18)22-25(35)29(39)32(42)30(40)26(22)36/h1-10H
InChI key
OXAQTDQMTJVZOI-UHFFFAOYSA-N
General description
Application
The field-effect hole and electron mobilities extracted from its single crystal ambipolar transistor are 7 × 10^-4 and 3 × 10^-3 cm^2 V^-1 s^-1 in the saturation regime. [2]
HOMO: -6.17 eV
LUMO: -4.00 eV
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service