Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

792217

Sigma-Aldrich

Titanyl phthalocyanine

type IV, >99%, Dye content

Synonym(s):

Oxo[29H,31H-phthalocyaninato(2-)]titanium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16N8OTi
CAS Number:
Molecular Weight:
576.39
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

>99%

form

powder (Blue)

composition

Dye content

mp

395.1 °C (dec.) (lit.)

λmax

700 nm

SMILES string

O=[Ti]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.O.Ti/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;;/h1-16H;;/q-2;;+2

InChI key

SJHHDDDGXWOYOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sensitizer for electrophotography
Sensitizer for electrophotography.

Legal Information

Product of Xerox®. Xerox® is a registered trademark of Xerox Corporation.
Xerox is a registered trademark of Xerox Corporation

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service