Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

792039

Sigma-Aldrich

Poly(L-lactide-co-5-methyl-5-allyloxycarbonyl-1,3-dioxan- 2-one)

10 mole % MAC, average Mn 40,000, PDI ≤1.9

Synonym(s):

PLA-co-MAC, PLLA-co-MAC, Poly(L-lactide-co-MAC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C3H4O2)n(C9H12O5)mH
UNSPSC Code:
12162002

form

powder

optical activity

[α]22/D -125.0°, c = 0.5% in chloroform

mol wt

average Mn 40,000

PDI

≤1.9

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

This biodegradeable random copolymer has pendant alkene functional groups along the backbone and a hydroxyl end-group. The pendant functionality along the chain enables bioconjugation of growth factors via thiol-ene click chemistry. The monomer ratio for MAC:L-lactide is 1:9.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service