Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

791601

Sigma-Aldrich

mIBX

Synonym(s):

1,3-Dihydro-1-hydroxy-3-oxo-1,2-benziodoxole-4-carboxylic acid 1-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5IO6
Molecular Weight:
324.03
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

reagent type: oxidant

functional group

carboxylic acid
iodo

storage temp.

−20°C

SMILES string

O=C(O1)C2=C(C(O)=O)C=CC=C2I1(O)=O

InChI

1S/C8H5IO6/c10-7(11)4-2-1-3-5-6(4)8(12)15-9(5,13)14/h1-3H,(H,10,11)(H,13,14)

InChI key

YGABLFBBILUMGY-UHFFFAOYSA-N

General description

mIBX (modified 2-iodoxybenzoic acid), a water-soluble hypervalent iodine compound, can be prepared from 3-nitrophthalic acid.[1] mIBX is an environment-friendly oxidizing agent that allows selective oxidation of allylic and benzylic alcohols in water.[2][1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and oxidation reactions of a user-and eco-friendly hypervalent iodine reagent.
Thottumkara AP and Vinod TK.
Tetrahedron Letters, 43(4), 569- 572 (2002)
Hypervalent iodine reagents for the oxidation of alcohols and their application to complex molecule synthesis.
Tohma H and Kita Y.
Advanced Synthesis & Catalysis, 346(2-3), 111-124 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service