791393
C106 Dye
≥85% (HPLC)
Synonym(s):
Greatcell Solar®, Ruthenate(2-), [[2,2′-bipyridine]-4,4′-dicarboxylato(2-)- N1,N1′][4,4′-bis[5-(hexylthio)-2-thienyl]-2,2′-bipyridine- N1,N1′]bis(thiocyanato-N)-, cis-Bis(isothiocyanato)(2,2′-bipyridyl-4,4′-dicarboxylato)( 4,4′-bis(5-(hexylthio)thiophen-2-yl)-2,2′-bipyridyl)ruthenium(II)
About This Item
Recommended Products
assay
≥85% (HPLC)
form
powder (black)
λmax
313 +/- 15 nm at 0.02 mM in DMF
348 +/- 15 nm at 0.02 mM in DMF
540 +/- 15 nm at 0.02 mM in DMF
SMILES string
OC(C1=CC(C2=CC(C(O)=O)=CC=N2)=NC=C1)=O.S=C=N[Ru]N=C=S.CCCCCCSC(S3)=CC=C3C4=CC(C5=CC(C6=CC=C(SCCCCCC)S6)=CC=N5)=NC=C4
InChI
1S/C30H36N2S4.C12H8N2O4.2CNS.Ru/c1-3-5-7-9-19-33-29-13-11-27(35-29)23-15-17-31-25(21-23)26-22-24(16-18-32-26)28-12-14-30(36-28)34-20-10-8-6-4-2;15-11(16)7-1-3-13-9(5-7)10-6-8(12(17)18)2-4-14-10;2*2-1-3;/h11-18,21-22H,3-10,19-20H2,1-2H3;1-6H,(H,15,16)(H,17,18);;;/q;;2*-1;+2
InChI key
ZEXAHKWZTIWSCT-UHFFFAOYSA-N
Application
Power conversion efficiencies of 11.7-12.1% achieved in high-efficiency dye-sensitized solar cells under air-mass 1.5G simulated sunlights.
Legal Information
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.
While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service