Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

791202

Sigma-Aldrich

3′-(Trifluoromethyl)-1,1′-biphenyl-2-carboxaldehyde

96%

Synonym(s):

2-[3-(Trifluoromethyl)phenyl]benzaldehyde, 3′-Trifluoromethylbiphenyl-2-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9F3O
CAS Number:
Molecular Weight:
250.22
UNSPSC Code:
12352100
NACRES:
NA.22

assay

96%

form

solid

refractive index

n20/D 1.551

density

1.281 g/mL at 25 °C

storage temp.

2-8°C

InChI

1S/C14H9F3O/c15-14(16,17)12-6-3-5-10(8-12)13-7-2-1-4-11(13)9-18/h1-9H

InChI key

STRFWILZQHGWOK-UHFFFAOYSA-N

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service