790915
Aluminum oxide
nanofiber, diam. × L <20 nm × 100 μm
Synonym(s):
Aluminum oxide, Alumina Nanofibers, Alumina nanowires, Aluminum oxide nanowires, Alumina
About This Item
form
nanofiber
diam. × L
<20 nm × 100 μm
mp
2040 °C (lit.)
>400 °C
SMILES string
O=[Al]O[Al]=O
InChI
1S/2Al.3O
Inchi Key
TWNQGVIAIRXVLR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service