Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

790869

Sigma-Aldrich

(S)-3,3′-Dibromo-1,1′-binaphthyl-2,2′-disulfonimide toluene complex

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
(C20H11Br2NO4S2)2 · C6H5CH3
Molecular Weight:
1198.63
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

optical activity

[α]22/D +133°, c = 0.5% in chloroform

reaction suitability

reaction type: Aldol Reaction
reaction type: Allylation
reaction type: Carbonyl/Imine Addition
reaction type: Friedel-Crafts Alkylation

mp

303-308 °C (decomposition without melting)

SMILES string

Cc1ccccc1.Brc2cc3ccccc3c4-c5c(c(Br)cc6ccccc56)S(=O)(=O)NS(=O)(=O)c24.Brc7cc8ccccc8c9-c%10c(c(Br)cc%11ccccc%10%11)S(=O)(=O)NS(=O)(=O)c79

InChI

1S/2C20H11Br2NO4S2.C7H8/c2*21-15-9-11-5-1-3-7-13(11)17-18-14-8-4-2-6-12(14)10-16(22)20(18)29(26,27)23-28(24,25)19(15)17;1-7-5-3-2-4-6-7/h2*1-10,23H;2-6H,1H3

InChI key

WQYRPPIFPJBANM-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Asymmetric counteranion-directed catalysis (ACDC) has a number of valuable applications in enantioselective synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service