Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

790850

Sigma-Aldrich

(S)-3,3′-Bis(4-trifluoromethylphenyl)-1,1′-binaphthyl-2,2′-disulfonimide

Synonym(s):

(11bS)-2,6-Bis[4-(trifluoromethyl)phenyl]dinaphtho[2,1-d:1′,2′-f][1,3,2]dithiazepine 3,3,5,5-tetraoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H19F6NO4S2
Molecular Weight:
683.64
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Aldol Reaction
reaction type: Allylation
reaction type: Carbonyl/Imine Addition
reaction type: Friedel-Crafts Alkylation

mp

340-360 °C

SMILES string

O=S(C1=C(C2=C(C=CC=C3)C3=CC(C4=CC=C(C(F)(F)F)C=C4)=C2S5(=O)=O)C6=C(C=CC=C6)C=C1C7=CC=C(C(F)(F)F)C=C7)(N5)=O

InChI

1S/C34H19F6NO4S2/c35-33(36,37)23-13-9-19(10-14-23)27-17-21-5-1-3-7-25(21)29-30-26-8-4-2-6-22(26)18-28(20-11-15-24(16-12-20)34(38,39)40)32(30)47(44,45)41-46(42,43)31(27)29/h1-18,41H

InChI key

FQLCMGCITBTGMR-UHFFFAOYSA-N

Application

Chiral Bronsted acid used in allylation of imines.

A Powerful Chiral Counteranion Motif for Asymmetric Catalysis

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Asymmetric counteranion-directed catalysis (ACDC) has a number of valuable applications in enantioselective synthesis.

Related Content

The List group focuses on the development of new concepts in catalysis. Since 1999, this research group has pioneered the development of organocatalysis as the third pillar of stereoselective catalysis, along side biocatalysis and transition metal catalysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service