Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

790222

Sigma-Aldrich

Polyglycolide-block-poly(ethylene glycol)-block-polyglycolide

PEG average Mn 400, PEG:glycolide 12:88

Synonym(s):

MODU-PRENE® SW1, PGA-PEG-PGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H2O2)m(C2H4O)nCH2CH2(C2H2O2)m
UNSPSC Code:
12352100

form

powder

feed ratio

PEG:glycolide 12:88

mol wt

PEG average Mn 400

transition temp

Tm >200 °C

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

This PGA-PEG-PGA triblock copolymer is a biodegradable, resorbable microparticulate modifier for use in drug delivery systems to induce the desired release profile.
Through combination of various gel formers, drug release profiles may be modulated to exhibit fast burst release through extended (up to 1 month) zero-order release profile.

Legal Information

Product of Poly-Med, Inc. Moduprene®
Moduprene is a registered trademark of Poly-Med, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service