Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

790001

Sigma-Aldrich

2′-Bromo-2,2,2-trifluoroacetophenone

97%

Synonym(s):

1-(2-Bromophenyl)-2,2,2-trifluoroethanone, 2-Bromo-α,α,α-trifluoroacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4BrF3O
CAS Number:
Molecular Weight:
253.02
Beilstein/REAXYS Number:
8312425
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.495

density

1.645 g/mL at 25 °C

functional group

bromo
fluoro
ketone

SMILES string

O=C(C(F)(F)F)C1=C(Br)C=CC=C1

InChI

1S/C8H4BrF3O/c9-6-4-2-1-3-5(6)7(13)8(10,11)12/h1-4H

InChI key

VNAWTGXXPNWISR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

214.0 °F - closed cup

flash_point_c

101.11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service