Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

779792

Sigma-Aldrich

β-Cyclodextrin-Citral complex

produced by Wacker Chemie AG, Burghausen, Germany, Wacker Chemie AG

Synonym(s):

Cavamax® W7-Citral complex

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

form

powder

manufacturer/tradename

Wacker Chemie AG

impurities

≤5 ppm heavy metals (USP)
≥8.5% citral content

loss

≤14.0% loss on drying

Legal Information

Cavamax is a registered trademark of Wacker Chemie AG

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service