Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

779768

Sigma-Aldrich

(S)-3,3′-Dibromo-2,2′-dimethoxy-1,1′-binaphthalene

≥98.0% (HPLC)

Synonym(s):

(S)-3,3′-Dibromo-2,2′-dimethoxy-1,1′-binaphthyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16Br2O2
CAS Number:
Molecular Weight:
472.17
Beilstein/REAXYS Number:
4712473
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥98.0% (HPLC)

optical purity

enantiomeric excess: ≥98.0%

SMILES string

COc1c(Br)cc2ccccc2c1-c3c(OC)c(Br)cc4ccccc34

InChI

1S/C22H16Br2O2/c1-25-21-17(23)11-13-7-3-5-9-15(13)19(21)20-16-10-6-4-8-14(16)12-18(24)22(20)26-2/h3-12H,1-2H3

Inchi Key

DFTUKDIMHCCQIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service