Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77914

Sigma-Aldrich

(S)-2-Aminohexane

≥97.0%

Synonym(s):

(S)-2-Hexylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15N
CAS Number:
Molecular Weight:
101.19
Beilstein/REAXYS Number:
1718994
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Quality Level

assay

≥97.0%

form

liquid

optical purity

enantiomeric ratio: >99.0:1.0 (GC)

SMILES string

CCCC[C@H](C)N

InChI

1S/C6H15N/c1-3-4-5-6(2)7/h6H,3-5,7H2,1-2H3/t6-/m0/s1

InChI key

WGBBUURBHXLGFM-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

26.6 °F

flash_point_c

-3 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service