Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77913

Sigma-Aldrich

(R)-(−)-1-Methoxy-2-propanol

≥98.5% (sum of enantiomers, GC)

Synonym(s):

(R)-(−)-Propylene glycol 1-methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O2
CAS Number:
Molecular Weight:
90.12
Beilstein/REAXYS Number:
1718941
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.5% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −22±2°, c = 10% in chloroform

refractive index

n20/D 1.403

bp

119-121 °C (lit.)

density

0.921 g/mL at 20 °C (lit.)

SMILES string

COC[C@@H](C)O

InChI

1S/C4H10O2/c1-4(5)3-6-2/h4-5H,3H2,1-2H3/t4-/m1/s1

InChI key

ARXJGSRGQADJSQ-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(-)-1-Methoxy-2-propanol is a chiral secondary alcohol. It is formed during the hydrolysis of (R)-1-methoxy-2-propyl-acetate in the presence of Candida antarctica lipase B.[1]

Application

(R)-(-)-1-Methoxy-2-propanol can be used as a reactant to prepare:
  • (S)-3-(ethoxycarbonyl)-5-((1-methoxypropan-2-yl)oxy)benzoic acid, a key intermediate to synthesize phenylethyl benzamide derivatives, which can be used as glucokinase activators.[2]
  • Chiral pyrazolopyrimidinone derivatives as potential phosphodiesterase enzyme (PDE5) inhibitors.[3]
  • Aryl or alkyl ethers via etherification reaction.[4]

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

91.4 °F - closed cup

flash_point_c

33 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly potent and selective chiral inhibitors of PDE5: an illustration of Pfeiffer?s rule
Bunnage ME, et al.
Bioorganic & medicinal chemistry letters, 18(23), 6033-6036 (2008)
Kinetic modeling of lipase catalyzed hydrolysis of (R/S)-1-methoxy-2-propyl-acetate as a model reaction for production of chiral secondary alcohols.
Berendsen WR, et al.
Journal of Biotechnology, 121(2), 213-226 (2006)
Nucleophilic Amination and Etherification of Aryl Alkyl Thioethers
Wang X, et al.
Organic Letters, 20(16), 4749-4753 (2018)
Kaapjoo Park et al.
Bioorganic & medicinal chemistry letters, 23(2), 537-542 (2012-12-12)
Novel benzamide derivatives were synthesized and tested at in vitro assay by measuring fold increase of glucokinase activity at 5.0 mM glucose concentration. Among the prepared compounds, YH-GKA was found to be an active glucokinase activator with EC(50) of 70

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service