Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

779083

Sigma-Aldrich

1-Azido-3-bromobenzene solution

~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):

3-Bromophenyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrN3
CAS Number:
Molecular Weight:
198.02
Beilstein/REAXYS Number:
2044137
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥95.0% (HPLC)

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

Brc1cccc(c1)N=[N+]=[N-]

InChI

1S/C6H4BrN3/c7-5-2-1-3-6(4-5)9-10-8/h1-4H

InChI key

KLKGUBBTVRHUGD-UHFFFAOYSA-N

Application

Review[1]

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-18.4 °F

flash_point_c

-28 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Bräse et al.
Angewandte Chemie (International ed. in English), 44(33), 5188-5240 (2005-08-16)
Since the discovery of organic azides by Peter Griess more than 140 years ago, numerous syntheses of these energy-rich molecules have been developed. In more recent times in particular, completely new perspectives have been developed for their use in peptide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service