Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

778931

Sigma-Aldrich

8-Phenyl-1-octanol

98% (GC)

Synonym(s):

8-Phenyloctyl alcohol, Benzeneoctanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22O
CAS Number:
Molecular Weight:
206.32
Beilstein/REAXYS Number:
2556146
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98% (GC)

form

liquid

refractive index

n20/D 1.504

density

0.942 g/mL at 20 °C

functional group

hydroxyl
phenyl

SMILES string

OCCCCCCCCc1ccccc1

InChI

1S/C14H22O/c15-13-9-4-2-1-3-6-10-14-11-7-5-8-12-14/h5,7-8,11-12,15H,1-4,6,9-10,13H2

InChI key

MGIGATOGROSKNW-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Kyung Song et al.
Journal of agricultural and food chemistry, 67(7), 2028-2035 (2019-01-31)
Caffeic acid phenethyl ester (CAPE) is an ester of a hydroxycinnamic acid (phenylpropanoid) and a phenylethanoid (2-phenylethanol; 2-PE), which has long been used in traditional medicine. Here, we synthesized 54 hydroxycinnamic acid-phenylethanoid esters by feeding 64 combinations of hydroxycinnamic acids

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service