Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

778680

Sigma-Aldrich

(2-Hydroxypropyl)-α-cyclodextrin

Wacker Chemie AG

Synonym(s):

Cavasol® W6 HP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201

form

powder

manufacturer/tradename

Wacker Chemie AG

extent of labeling

molar substitution (report result)

impurities

propylene glycol (report result)

loss

loss on drying (report result)

mp

245 °C (dec.) (lit.)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Cavasol is a registered trademark of Wacker Chemie AG

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel J Smith et al.
The European journal of neuroscience, 30(3), 493-503 (2009-08-07)
The orexin/hypocretin system has recently been implicated in reward-processing and addiction. We examined the involvement of the orexin system in cue-induced reinstatement of extinguished cocaine-seeking by administering the orexin 1 receptor antagonist SB-334867 (SB) or the orexin 2 receptor antagonist
D E Rusyniak et al.
Neuroscience, 154(4), 1619-1626 (2008-06-07)
When given in a warm environment MDMA (3,4-methylenedioxymethamphetamine, ecstasy) causes hyperthermia by increasing interscapular brown adipose tissue (iBAT) heat production and decreasing heat loss via cutaneous vasoconstriction. When given in a cold environment, however, MDMA causes hypothermia by an unknown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service