Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

778486

Sigma-Aldrich

Benzoyl azide solution

~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):

Benzazide solution, Benzoic acid azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O
CAS Number:
Molecular Weight:
147.13
Beilstein/REAXYS Number:
637869
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥95.0% (HPLC)

form

solution

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

[N-]=[N+]=NC(=O)c1ccccc1

InChI

1S/C7H5N3O/c8-10-9-7(11)6-4-2-1-3-5-6/h1-5H

InChI key

PJHUABJTDFXYRQ-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-27.4 °F

flash_point_c

-33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service