Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

778230

Sigma-Aldrich

Bis(triisopropylcyclopentadienyl)barium tetrahydrofuran adduct

98%

Synonym(s):

Bis(1,2,4-triisopropylcyclopentadienyl)barium, Bis[1,2,4-tris(1-methylethyl)-2,4-cyclopentadien-1-yl]barium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ba(C5(C3H7)3H2)2] ⋅ 2(C4H8O)
UNSPSC Code:
12352300
PubChem Substance ID:

assay

98%

form

powder

reaction suitability

core: barium

mp

284-289 °C

SMILES string

CC(C)[C]1[C][C](C(C)C)[C][C]1C(C)C.CC(C)[C]2[C][C](C(C)C)[C][C]2C(C)C.[Ba].C3CCOC3.C4CCOC4

InChI

1S/2C14H23.2C4H8O.Ba.2H/c2*1-9(2)12-7-13(10(3)4)14(8-12)11(5)6;2*1-2-4-5-3-1;;;/h2*7-11H,1-6H3;2*1-4H2;;;

InChI key

XPFYNIUESMDERR-UHFFFAOYSA-N

Application

Cyclopentadienyl compounds of Barium are very commonly used ALD/CVD precursors for depositing Barium containing thin films; example BaTiO3; BaZrO3
films. Cyclopentadienyl groups form weaker bonds with Barium, but have stronger bonds within the ligand thus preventing carbon contamination of the
films. Cyclopentadienyl precursors of barium sublime under reduced pressures, with tetrahydrofuran adducts showing volatility also under atmospheric
pressure. The tetrahydrofuran adducts loose the coordinated THF when evaporated. Complexes with bulky cyclopentadienyl ligands are more thermally
stable and volatile.
Barium containing thin films finds applications as host lattices for luminescent materials, high temperature superconductors, high permittivity
dielectrics and ferroelectrics.

pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vehkamaki; M.; Hatanpaa; T.; Hanninen; T.; Ritala; M.; Leskela; M.
Electrochemical and Solid-State Letters, 2, 504-504 (1999)
Holme; T. P.; Prinz; F. B.
The Journal of Physical Chemistry, 111, 8147-8147 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service