Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

778060

Sigma-Aldrich

4,4,4-Trifluorocrotonic acid

96%

Synonym(s):

4,4,4-Trifluoro-2-butenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3F3O2
CAS Number:
Molecular Weight:
140.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

mp

52-55 °C

SMILES string

OC(/C=C/C(F)(F)F)=O

InChI

1S/C4H3F3O2/c5-4(6,7)2-1-3(8)9/h1-2H,(H,8,9)/b2-1+

InChI key

QZBAYURFHCTXOJ-OWOJBTEDSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

172.0 °F - closed cup

flash_point_c

77.77 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service