Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

777811

Sigma-Aldrich

3,5-Dibromopyridine-4-methanol

97%

Synonym(s):

3,5-Dibromo-4-hydroxymethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Br2NO
CAS Number:
Molecular Weight:
266.92
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

108-110 °C

functional group

bromo
hydroxyl

SMILES string

OCc1c(Br)cncc1Br

InChI

1S/C6H5Br2NO/c7-5-1-9-2-6(8)4(5)3-10/h1-2,10H,3H2

InChI key

ZHNPEZCPMQFETF-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service