Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

777587

Sigma-Aldrich

4-Amino-3-hydroxypiperidine

95%

Synonym(s):

3-Hydroxy-4-aminopiperidine, 4-Amino-3-piperidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2O
CAS Number:
Molecular Weight:
116.16
UNSPSC Code:
12352100
NACRES:
NA.22

assay

95%

form

solid

mp

70-75 °C

storage temp.

2-8°C

InChI

1S/C5H12N2O/c6-4-1-2-7-3-5(4)8/h4-5,7-8H,1-3,6H2

InChI key

UVBKPWNFXFUDOU-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service