Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

777277

Sigma-Aldrich

1-(Phthalimidomethyl) 2-bromoisobutyrate

97%

Synonym(s):

ATRP initiator, protected amine BIB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12BrNO4
CAS Number:
Molecular Weight:
326.14
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

solid

mp

89-94 °C

storage temp.

2-8°C

SMILES string

CC(C)(Br)C(=O)OCN1C(=O)c2ccccc2C1=O

InChI

1S/C13H12BrNO4/c1-13(2,14)12(18)19-7-15-10(16)8-5-3-4-6-9(8)11(15)17/h3-6H,7H2,1-2H3

InChI key

AIPGWKMNUNTLCB-UHFFFAOYSA-N

Application

This ATRP initiator can be used to generate a polymer with a protected amine end group functionality.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Schroder; Kristin;
ACS Macro Letters, 1, 1037-1040 (2012)
Harry C Pappas et al.
ACS applied materials & interfaces, 7(50), 27632-27638 (2015-11-26)
Mitigation of bacterial adhesion and subsequent biofilm formation is quickly becoming a strategy for the prevention of hospital-acquired infections. We demonstrate a basic strategy for surface modification that combines the ability to control attachment by microbes with the ability to
Stefanie L Baker et al.
Biomacromolecules, 19(9), 3798-3813 (2018-08-08)
The power and elegance of protein-polymer conjugates has solved many vexing problems for society. Rational design of these complex covalent hybrids depends on a deep understanding of how polymer physicochemical properties impact the conjugate structure-function-dynamic relationships. We have generated a
Xiujun Ren et al.
Journal of chromatography. A, 1618, 460904-460904 (2020-01-30)
A novel chiral stationary phase (CSP) was prepared through the reaction of surface-initiated atom transfer radical polymerization (ATRP) by the copolymerization of thermoresponsive N-isopropylacrylamide (NIPAM) and β-cyclodextrin (β-CD) on the silica beads for high performance liquid chromatography (HPLC). X-ray photoelectron

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service