Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

777153

Sigma-Aldrich

3-Bromocarbazole

97% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8BrN
CAS Number:
Molecular Weight:
246.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97% (GC)

form

solid

mp

196-201 °C

functional group

bromo

SMILES string

BrC1=CC2=C(C=C1)NC3=C2C=CC=C3

InChI

1S/C12H8BrN/c13-8-5-6-12-10(7-8)9-3-1-2-4-11(9)14-12/h1-7,14H

InChI key

LTBWKAYPXIIVPC-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Mumbo et al.
Environmental science and pollution research international, 24(35), 27525-27538 (2017-10-06)
Bromocarbazoles and chlorocarbazoles are emerging environmental contaminants that have been reported to be persistent and possessing dioxin-like toxicity; however, their photodegradative fate in water is unknown. The photodegradation of 3-bromocarbazole, 3-chlorocarbazole, and 3,6-dichlorocarbazole was determined in ultrapure water. They proceeded
Jiehong Guo et al.
Environmental science & technology, 51(1), 89-97 (2016-12-21)
Polyhalogenated carbazoles (PHCZs) have been increasingly detected in the environment. Their similarities in chemical structure with legacy pollutants and their potential toxicity have caused increasing concern. In this work, 112 Ponar grab and 28 core sediment samples were collected from
Wen-Ling Chen et al.
Chemosphere, 150, 586-595 (2016-02-28)
Carbazole and some of its derivatives may possess dioxin-like toxicity and could be persistent in the environment, but information on their distribution and environmental fate is limited. This study developed and validated an ultra-trace targeted-analysis method for the determination of
Li Fang et al.
Ecotoxicology and environmental safety, 184, 109609-109609 (2019-09-14)
Carbazole and halogenated carbazoles have been widely detected throughout the environment in soil, river deposits, and lake sediments. Human exposure to these compounds may occur through inhalation, drinking water, dietary intake and/or skin contact, and exposure levels in the body
Yan Wu et al.
Environmental science & technology, 52(8), 4536-4545 (2018-03-23)
Polyhalogenated carbazoles (PHCZs) were recently discovered in Great Lakes sediment and other aquatic systems. However, knowledge about their bioaccumulation and potential risks to fish and wildlife remains very limited. The present study investigated PHCZs in Great Lakes lake trout (

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service