Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

776866

Sigma-Aldrich

Poly(maleic anhydride-alt-1-octadecene)

high viscosity

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

viscosity

≤2,000 cP

mp

>400 °C

transition temp

Tm 115-135 °C
softening point 120-130 °C

density

0.97 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Release coatings, anti-corrosive, thickening agent, chelant/surfactant and cross-linker for epoxy or alcohol functional polymers.

Features and Benefits

Readily derivatized to acid salt, amide, ester or imide. Alternating polymer.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service