Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

776831

Sigma-Aldrich

2-Chloro-3-hydroxypropionic acid

95%

Synonym(s):

2-Chloro-3-hydroxypropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5ClO3
CAS Number:
Molecular Weight:
124.52
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

52-56 °C

functional group

carboxylic acid
chloro
hydroxyl

storage temp.

2-8°C

SMILES string

OCC(C(O)=O)Cl

InChI

1S/C3H5ClO3/c4-2(1-5)3(6)7/h2,5H,1H2,(H,6,7)

InChI key

GAHMZBZGINHHJR-UHFFFAOYSA-N

pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service