Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

776815

Sigma-Aldrich

Diethylene glycol butyl ether methacrylate

contains <200 ppm monomethyl ether hydroquinone as inhibitor, 97% (GC)

Synonym(s):

Butyl PEG methacrylate, DEGBEMA, PEGBEMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O4
CAS Number:
Molecular Weight:
230.30
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97% (GC)

form

liquid

contains

<200 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.443

density

0.963 g/mL at 25 °C

storage temp.

−20°C

SMILES string

C=C(C)C(OCCOCCOCCCC)=O

InChI

1S/C12H22O4/c1-4-5-6-14-7-8-15-9-10-16-12(13)11(2)3/h2,4-10H2,1,3H3

InChI key

QGTBRAFPWNISIJ-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

276.8 °F

flash_point_c

136 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service