Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

776378

Sigma-Aldrich

Poly(L-lactide), amine terminated

average Mn 2,500, PDI ≤1.3

Synonym(s):

Amino-functionalized polylactide, NH2 PLLA, PLA-NH2, amino-PLLA, aminoPLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H9NO(C3H4O2)n
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

optical activity

[α]22/D -131°, c = 0.5% in chloroform

mol wt

average Mn 2,500

PDI

≤1.3

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Drug Delivery including coating nanoparticles[1]; end-group functionalization[2]; macroinitiator or block precursor.[3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly(lactide)-block-Poly(HEMA) Block Copolymers: An Orthogonal One-Pot Combination of ROP and ATRP, Using a Bifunctional Initiator
Wolf F F, et al.
Macromolecules, 42(15), 5622-5628 (2009)
Synthesis of Shape Amphiphiles Based on Functional Polyhedral Oligomeric Silsesquioxane End-Capped Poly(L-Lactide) with Diverse Head Surface Chemistry
Zhang W B, et al.
Macromolecules, 44(8), 2589-2596 (2011)
Preparation of Protein-Loaded Poly(L-Lactide) Microspheres by Solution-Enhanced Dispersion by Supercritical CO2
Chen A Z, et al.
Journal of Biomimetics, Biomaterials, and Tissue Engineering, 11, 93-100 (2011)

Articles

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service