Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775932

Sigma-Aldrich

[Ru(p-cymene)(O-DPPBS)Cl]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28ClO3PRuS
CAS Number:
Molecular Weight:
612.08
UNSPSC Code:
12352300
NACRES:
NA.22

reaction suitability

core: ruthenium
reagent type: catalyst

mp

233-242 °C (decomposition)

Application

Catalyst has been employed in the reductive amination of allylic alcohols.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeyneb Sahli et al.
Organic letters, 13(15), 3964-3967 (2011-06-28)
Straighforward access to various saturated amines from allylic alcohols and isostructural mixture can now be achieved in the presence of arene ruthenium catalyst featuring phosphinesulfonate ligand and a hydrogen donor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service