Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775894

Sigma-Aldrich

[Ru(p-cymene)(O-tBuPPBS)Cl]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32ClO3PRuS
CAS Number:
Molecular Weight:
592.09
UNSPSC Code:
12161600
NACRES:
NA.22

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

149-156 °C (decomposition)

Application

Catalyst for N-alkylation of amines with alcohols

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Synthesis & Catalysis, 352, 3141-3146 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service