Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775886

Sigma-Aldrich

2-(4-Methoxyphenylcarbonothioylthio)ethanoic acid

≥97%

Synonym(s):

(4-Methoxybenzothioylthio)ethanoic acid, 2-[(4-Methoxyphenylthioxomethyl)thio]ethanoic acid, CTA, RAFT dithiobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O3S2
CAS Number:
Molecular Weight:
242.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

assay

≥97%

form

liquid

mp

122-125 °C

Storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(=S)SCC(O)=O

InChI

1S/C10H10O3S2/c1-13-8-4-2-7(3-5-8)10(14)15-6-9(11)12/h2-5H,6H2,1H3,(H,11,12)

Inchi Key

OYGMVZNPQIMDSA-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization; especially suited for the polymerization of methacrylate and methacrylamide monomers. Chain Transfer Agent (CTA) with electron donating group to increase fragmentation and reduce polymerization time.

Packaging

Amber glass bottle; light sensitive. Handle under nitrogen

Pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Articles

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service