Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775878

Sigma-Aldrich

3,6-Bis(3-ethyl-N-carbazolyl)-N-phenylcarbazole

99% (HPLC)

Synonym(s):

3,3′′-Diethyl-9′-phenyl-9′H-9,3′:6′,9′′-tercarbazole, 3,3′′-Diylbis(ethyl)-9′-phenyl-9,3′: 6′,9′′-ter-9H-carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H35N3
Molecular Weight:
629.79
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

99% (HPLC)

form

powder

mp

205-220 °C

λmax

241 nm in dichloromethane

fluorescence

λem 521 nm (λmax)

SMILES string

CCC(C=C1)=CC2=C1N(C3=CC4=C(C=C3)N(C5=CC=CC=C5)C6=C4C=C(N7C8=C(C=C(CC)C=C8)C9=C7C=CC=C9)C=C6)C%10=C2C=CC=C%10

InChI

1S/C46H35N3/c1-3-30-18-22-43-37(26-30)35-14-8-10-16-41(35)48(43)33-20-24-45-39(28-33)40-29-34(21-25-46(40)47(45)32-12-6-5-7-13-32)49-42-17-11-9-15-36(42)38-27-31(4-2)19-23-44(38)49/h5-29H,3-4H2,1-2H3

InChI key

LMKCXAKDJFQWMK-UHFFFAOYSA-N

Application

Host materials for efficient solution-processed red and green phosphorescent devices

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service