Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775754

Sigma-Aldrich

4-Hydroxy-6-methyl-2-methylthiopyrimidine

97%

Synonym(s):

6-Methyl-2-(methylthio)-1H-pyrimidin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2OS
CAS Number:
Molecular Weight:
156.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

224-226 °C

functional group

thioether

SMILES string

CSc1nc(C)cc(O)n1

InChI

1S/C6H8N2OS/c1-4-3-5(9)8-6(7-4)10-2/h3H,1-2H3,(H,7,8,9)

InChI key

OCOHPSHRDKBGOZ-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service