Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775436

Sigma-Aldrich

Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]{[5-(2-ethoxy-2-oxoethanamido)]-2-isopropoxybenzylidene}ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H43Cl2N3O4Ru
CAS Number:
Molecular Weight:
741.71
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

104-152 °C (decomposition)

storage temp.

2-8°C

SMILES string

CC1=CC(C)=CC(C)=C1N(CCN2C3=C(C)C=C(C)C=C3C)C2=[Ru](Cl)(Cl)=CC4=C(OC(C)C)C=CC(NC(C(OCC)=O)=O)=C4

InChI

1S/C21H26N2.C14H17NO4.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-5-18-14(17)13(16)15-11-6-7-12(10(4)8-11)19-9(2)3;;;/h9-12H,7-8H2,1-6H3;4,6-9H,5H2,1-3H3,(H,15,16);2*1H;/q;;;;+2/p-2

InChI key

JILXLQWXSILBOS-UHFFFAOYSA-L

Legal Information

Umicore M7 catalysts are sold in cooperation with Umicore and OmegaCat Systems for research purposes only.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service