Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775320

Sigma-Aldrich

Grubbs Catalyst® M520

Synonym(s):

Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]{[2-(2-oxopropoxy)phenyl]methylene}ruthenium(II), Grubbs Catalyst® M52

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H36Cl2N2O2Ru
CAS Number:
Molecular Weight:
640.61
UNSPSC Code:
12352300
NACRES:
NA.22

mp

184-189 °C

storage temp.

2-8°C

SMILES string

CC1=C(C(C)=CC(C)=C1)n2c([Ru]34(Cl)(Cl)=[C]C5=C(C=CC=C5)O3C(C)C(C)=O4)n(C(C(C)=CC(C)=C6)=C6C)CC2

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 2 - Ox. Sol. 2 - Skin Irrit. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service