Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775185

Sigma-Aldrich

Ethyl 4,4-difluoroacetoacetate

96% (mixture of tautomers)

Synonym(s):

4,4-Difluoro-3-oxobutanoic acid ethyl ester, 4,4-Difluoro-3-oxobutyric acid ethyl ester, 4,4-Difluoroacetoacetic acid ethyl ester, Ethyl 4,4-difluoro-3-ketobutyrate, Ethyl 4,4-difluoro-3-oxobutanoate, Ethyl 4,4-difluoro-3-oxobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8F2O3
CAS Number:
Molecular Weight:
166.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

96% (mixture of tautomers)

form

liquid

refractive index

n20/D 1.409

density

1.269 g/mL at 25 °C

SMILES string

CCOC(=O)CC(=O)C(F)F

InChI

1S/C6H8F2O3/c1-2-11-5(10)3-4(9)6(7)8/h6H,2-3H2,1H3

InChI key

CBDPWKVOPADMJC-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

156.0 °F

flash_point_c

68.88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service