Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

775088

Sigma-Aldrich

Sodium bis(trimethylsilyl)amide solution

1.0 M in 2-methyltetrahydrofuran

Synonym(s):

Hexamethyldisilazane sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NNa
CAS Number:
Molecular Weight:
183.37
Beilstein/REAXYS Number:
3629917
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

form

liquid

concentration

1.0 M in 2-methyltetrahydrofuran

density

0.876 g/mL at 25 °C

SMILES string

C[Si](C)(C)N([Na])[Si](C)(C)C

InChI

1S/C6H18NSi2.Na/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

WRIKHQLVHPKCJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Strong organic base in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

10.4 °F

flash_point_c

-12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nur Hazlin Hazrin-Chong et al.
Journal of microbiological methods, 90(2), 96-99 (2012-05-09)
The use of hexamethyldisilazane (HMDS) as a drying agent was investigated in the specimen preparation for scanning electron microscopy (SEM) imaging of bacterial surface colonization on sub-bituminous coal. The ability of microbes to biofragment, ferment and generate methane from coal
Nan Li et al.
Lab on a chip, 8(12), 2105-2112 (2008-11-22)
High-density live cell array serves as a valuable tool for the development of high-throughput immunophenotyping systems and cell-based biosensors. In this paper, we have, for the first time, demonstrated a simple fabrication process to form the hexamethyldisilazane (HMDS) and poly(ethylene
Elsa Vennat et al.
Dental materials : official publication of the Academy of Dental Materials, 25(6), 729-735 (2009-01-29)
The objectives of this study were to assess demineralized dentin porosity and quantify the different porous features distribution within the material using mercury intrusion porosimetry (MIP) technique. We compared hexamethyldisilazane (HMDS) drying and lyophilization (LYO) (freeze-drying) in sample preparation. Fifty-six
Xiaoguang Li et al.
Chemical communications (Cambridge, England), 47(38), 10761-10763 (2011-08-30)
A silica microsphere suspension and a silica sol are employed in a two-step dipping process for the preparation of a superhydrophobic surface. It's not only a facile way to achieve the lotus effect, but can also create a multi-functional surface
Juliana Tsz Yan Lee et al.
Scanning, 34(1), 12-25 (2012-04-26)
Common dehydration methods of cells on biomaterials for scanning electron microscopy (SEM) include air drying, hexamethyldisilazane (HMDS) or tetramethysilane (TMS) treatment and critical point drying (CPD). On the other side, freeze-drying has been widely employed in dehydrating biological samples and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service