774685
1-Dodecylmagnesium bromide solution
0.5 M in 2-methyltetrahydrofuran
Synonym(s):
Dodecylmagnesium bromide solution
About This Item
Recommended Products
form
solution
reaction suitability
reaction type: Grignard Reaction
concentration
0.5 M in 2-methyltetrahydrofuran
density
0.908 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCC[Mg]Br
InChI
1S/C12H25.BrH.Mg/c1-3-5-7-9-11-12-10-8-6-4-2;;/h1,3-12H2,2H3;1H;/q;;+1/p-1
Inchi Key
JSDNASSZWGNUBO-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry
Packaging
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 2
supp_hazards
Storage Class
4.2 - Pyrophoric and self-heating hazardous materials
wgk_germany
WGK 3
flash_point_f
6.8 °F
flash_point_c
-14 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service