Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

774383

Sigma-Aldrich

Diethyl 3-(trifluoromethyl)benzylphosphonate

97%

Synonym(s):

{[3-(Trifluoromethyl)phenyl]methyl}phosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16F3O3P
CAS Number:
Molecular Weight:
296.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.455

bp

282-296 °C/760 mmHg

density

1.223 g/mL at 25 °C

functional group

fluoro
phosphonate

SMILES string

CCOP(=O)(Cc1cccc(c1)C(F)(F)F)OCC

InChI

1S/C12H16F3O3P/c1-3-17-19(16,18-4-2)9-10-6-5-7-11(8-10)12(13,14)15/h5-8H,3-4,9H2,1-2H3

InChI key

JRANUXHJUGOHIM-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service