774278
Diethyl sulfite
98%
Synonym(s):
DES, Ethyl sulfite
About This Item
Recommended Products
Quality Level
assay
98%
form
liquid
refractive index
n20/D 1.414 (lit.)
n20/D 1.414
bp
158-160 °C (lit.)
density
1.077 g/mL at 25 °C (lit.)
1.078 g/mL at 25 °C
application(s)
battery manufacturing
SMILES string
CCOS(=O)OCC
InChI
1S/C4H10O3S/c1-3-6-8(5)7-4-2/h3-4H2,1-2H3
InChI key
NVJBFARDFTXOTO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Warning
hcodes
pcodes
Hazard Classifications
Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
125.6 °F - closed cup
flash_point_c
52 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service