Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

774219

Sigma-Aldrich

5-Bromothiophene-3-carboxaldehyde

98%

Synonym(s):

5-Bromo-3-thiophenecarbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrOS
CAS Number:
Molecular Weight:
191.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.624

bp

237-242 °C

density

1.757 g/mL at 25 °C

functional group

aldehyde
bromo

storage temp.

2-8°C

SMILES string

Brc1cc(C=O)cs1

InChI

1S/C5H3BrOS/c6-5-1-4(2-7)3-8-5/h1-3H

InChI key

ZKLBPUYMTHPNOQ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service