Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

773794

Sigma-Aldrich

4-Aminophenyl propargyl ether

95% (HPLC)

Synonym(s):

4-(prop-2-ynyloxy)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

95% (HPLC)

form

solid

mp

47-53 °C

storage temp.

2-8°C

SMILES string

Nc1ccc(OCC#C)cc1

InChI

1S/C9H9NO/c1-2-7-11-9-5-3-8(10)4-6-9/h1,3-6H,7,10H2

InChI key

FPLSLCJCSKFAMA-UHFFFAOYSA-N

Application

This monomer has amino functionality as well as acetylene functionality for copper mediated ligation or functionalization. This can be used to copolymerize with maleic anhydrides and maleimides.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service