Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

773778

Sigma-Aldrich

2-(Phenylcarbonothioylthio)propanoic acid

98%

Synonym(s):

2-[(Phenylthioxomethyl)thio]propanoic acid, Benzodithioate RAFT, Dithiobenzoate 2-propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2S2
CAS Number:
Molecular Weight:
226.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Quality Level

assay

98%

form

solid

mp

81-88 °C

Storage temp.

2-8°C

SMILES string

CC(SC(=S)c1ccccc1)C(O)=O

InChI

1S/C10H10O2S2/c1-7(9(11)12)14-10(13)8-5-3-2-4-6-8/h2-7H,1H3,(H,11,12)

Inchi Key

HPSQKMAZWIIJBT-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization; Chain Transfer Agent (CTA)
Reversible Addition Fragmentation Chain Transfer (RAFT) Polymerization

Packaging

Amber glass bottle; please keep out of light and pack under N2.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service