Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

773522

Sigma-Aldrich

tert-Butylmagnesium chloride solution

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CMgCl
CAS Number:
Molecular Weight:
116.87
Beilstein/REAXYS Number:
3535403
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solution

reaction suitability

reaction type: Grignard Reaction

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

density

0.878 g/mL at 25 °C

greener alternative category

SMILES string

CC(C)(C)[Mg]Cl

InChI

1S/C4H9.ClH.Mg/c1-4(2)3;;/h1-3H3;1H;/q;;+1/p-1

InChI key

ZDRJSYVHDMFHSC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product, 1.0 M in 2-methyltetrahydrofuran aligns with "Safer Solvents and Auxiliaries", "Use of Renewable Feedstocks" and "Inherently Safer Chemistry for Accident Prevention". Find details here.

Application

Grignard reagent has been utilized in nucleophilic additions and substitutions; metal-halogen exchanges and Kumada couplings.
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF)

2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

-22.0 °F

flash_point_c

-30 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service