Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

773514

Sigma-Aldrich

PBDT(EH)-TPD(Oct)

Synonym(s):

PBDTTPD, Poly[(5,6-dihydro-5-octyl-4,6-dioxo-4H-thieno[3,4-c]pyrrole-1,3-diyl)[4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl]]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C40H53NO4S3)n
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

mp

>300 °C

λmax

627 nm (thin film)

orbital energy

 1.83 eV (opt)
HOMO 5.4 eV 
LUMO 3.75 eV 

P-typeOFET device performance

Mobility

    P-type
  • Mobility: 2 x 10^-4 cm2/V·s

OPV device performance

ITO/PEDOT:PSS/ PBDTTPD:fullerene (1:1.5 by weight)/Ca/AlAdv. Energy Mater. 2012, 3, 220

  • Short-circuit current density (Jsc): 11.1 mA/cm2
  • Open-circuit voltage (Voc): 0.89 V
  • Fill Factor (FF): 60
  • Power Conversion Efficiency (PCE): 6 %

storage temp.

2-8°C

General description

PBDT(EH)-TPD(Oct) is a conducting polymer with benzothiophene as the electron donor and thieno[3,4-c]pyrrole-4,6-dione as the electron withdrawing units. It can be used to form a π-conjugating system for the development of a variety of high performance electrochemical devices.[1][2]

application

PBDT(EH)-TPD(Oct) is a semiconducting polymer which can be used in the fabrication of polymeric solar cells and photovoltaic cells with high power conversion efficiency.[1][2][3][4][5]
Use in organic high efficiency photovoltaic cells (PCE 6.8%) and field effect transistors.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dependence of crystallite formation and preferential backbone orientations on the side chain pattern in PBDTTPD polymers
El Labban A, et al.
ACS Applied Materials & Interfaces, 6(22), 19477-19481 (2014)
Intensity dependent femtosecond dynamics in a PBDTTPD-based solar cell material
Paraecattil AA, et al.
The Journal of Physical Chemistry Letters, 3(20), 2952-2958 (2012)
Advances in Functional Materials, 21(4), 718-728 (2011)
Ring substituents mediate the morphology of PBDTTPD-PCBM bulk-heterojunction solar cells
Warnan J, et al.
Chemistry of Materials, 26(7), 2299-2306 (2014)
Efficient polymer solar cells based on the copolymers of benzodithiophene and thienopyrroledione
Zhang Y, et al.
Chemistry of Materials, 22(9), 2696-2698 (2010)

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service