Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

773387

Sigma-Aldrich

Poly(2-ethyl-2-oxazoline) α-methyl, ω-2-hydroxyethylamine terminated

average Mn 10,000, PDI <1.3

Synonym(s):

POZ, Poly(2-ethyl-2-oxazoline) hydroxyethyl amine, amino terminated polyoxazoline, aminoPEOx, polyoxazoline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C5H9NO)nNHC2H4OH
UNSPSC Code:
12162002
NACRES:
NA.23

description

Tg 59°C

form

solid

mol wt

average Mn 10,000

PDI

<1.3

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Amine polyoxazolines can be covalently attached through conjugation to carboxyl groups or sulfhydryl groups. Amines can also attach to oxidized carbohydrate groups (usually aldehydes) by reductive amination. Polyoxazolines are a biocompatible, hydrophilic, amorphous alternatives to poly(ethylene glycol).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Poly(2-oxazoline)s, synthesized via LCROP, resemble polypeptides and have various applications in polymer chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service