Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

773328

Sigma-Aldrich

Poly(vinyl acetate), cyanomethyl diphenylcarbamodithioate

average Mn 5,000, PDI <1.2

Synonym(s):

PVA, PVA-RAFT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H2N(C4H6O2)nC13H10NS2
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

mol wt

average Mn 5,000

transition temp

Tm 288-293 °C

PDI

<1.2

General description

All polyethylene imine polymers are hydrophilic and may contain approx. 30% hydrated water.

Application

Poly(vinyl acetate) is commonly used in biomedical applications. The end group functionality contains a cyanomethyl group; and the RAFT agent diphenylcarbamodithioate which will allow for additional polymerization to generate a diblock copolymer. This RAFT agent is well-suited for vinyl acetates and vinyl benzoate

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akin Akinc et al.
The journal of gene medicine, 7(5), 657-663 (2004-11-16)
The relatively high transfection efficiency of polyethylenimine (PEI) vectors has been hypothesized to be due to their ability to avoid trafficking to degradative lysosomes. According to the proton sponge hypothesis, the buffering capacity of PEI leads to osmotic swelling and
Godbey, W.T.:
J. Controlled Release, 60(2-3), 149-160 (1999)

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service