Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

773190

Sigma-Aldrich

Fmoc-N-Me-Thr(tBu)-OH

97%

Synonym(s):

Fmoc-N-methyl-O-tert-butyl-L-threonine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29NO5
CAS Number:
Molecular Weight:
411.49
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

assay

97%

form

powder or crystals

optical activity

[α]22/D +16.0°, c = 0.5% in methanol

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

220-225 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

InChI

1S/C24H29NO5/c1-15(30-24(2,3)4)21(22(26)27)25(5)23(28)29-14-20-18-12-8-6-10-16(18)17-11-7-9-13-19(17)20/h6-13,15,20-21H,14H2,1-5H3,(H,26,27)/t15-,21+/m1/s1

InChI key

VIUVLZHFMIFLHU-VFNWGFHPSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service