Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

773034

Sigma-Aldrich

Potassium (3-butenyl)trifluoroborate

90%

Synonym(s):

Potassium homoallyltrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7BF3K
CAS Number:
Molecular Weight:
162.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

90%

form

solid

mp

>300 °C

SMILES string

[K+].F[B-](F)(F)CCC=C

InChI

1S/C4H7BF3.K/c1-2-3-4-5(6,7)8;/h2H,1,3-4H2;/q-1;+1

InChI key

UPXWGXKSNOQOKH-UHFFFAOYSA-N

Application

Organotrifluoroborate involved in:
  • Catalytic allylboration[1]
  • Stereoselective nucleophilic addition[2]
  • Pd-catalyzed heterocyclizations[3]
  • Oxidation reactions[4] and Oxidative Mannich reactions[5]
  • Cross-coupling reactions[6]


Organotrifluoroborates as versatile and stable boronic acid surrogates

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation of alkyl trifluoroborates: an opportunity for tin-free radical chemistry.
Geoffroy Sorin et al.
Angewandte Chemie (International ed. in English), 49(46), 8721-8723 (2010-10-12)
The development of a Selectfluor-mediated oxidative Mannich reaction
M. H. Daniels and J. Hubbs,
Tetrahedron Letters, 52, 3543-3546 (2011)
Catalytic activation of pinacolyl allylboronate with indium(I): development of a general catalytic allylboration of ketones.
Uwe Schneider et al.
Angewandte Chemie (International ed. in English), 46(31), 5909-5912 (2007-06-20)
Michael J Burns et al.
Dalton transactions (Cambridge, England : 2003), 39(43), 10391-10400 (2010-10-15)
A new synthetic methodology for the catalytic C-H functionalisation of 2-pyrones is described which proceeds regioselectively at the C3 position, mirroring the observed regioselectivity in 6π-electrocyclisation/oxidative aromatisation reactions of related compounds. Insight into the reaction mechanism is provided, with support
A new organic transformation by introducing crotyl/allyltrifluoroborates in cross-coupling reaction with aroyl chlorides
M. Al-Masum and K. Liu,
Tetrahedron Letters, 52, 5090-5093 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service